Merck
  • A Comparison of ACQ, AIE and AEE-Based Polymers Loaded on Polyurethane Foams as Sensors for Explosives Detection.
A Comparison of ACQ, AIE and AEE-Based Polymers Loaded on Polyurethane Foams as Sensors for Explosives Detection.
Sensors (Basel, Switzerland) (2018-05-16)
Zhiwei Chu, Zhuxin Fan, Xiang Zhang, Xiaofeng Tan, Dongxu Li, Guohua Chen, Qinghua Zhao
ABSTRACT
An aggregation-caused quenching (ACQ)-active polymer (PF), an aggregation-induced emission (AIE)-active polymer (PFTPE) and an aggregation-enhanced emission (AEE)-active polymer (PTTPE) were synthesized by tetraphenylethane (TPE), fluorene and thiophene moieties. Polyurethane (PU) foams modified by PF, PFTPE and PTTPE, namely PU-PF, PU-PFTPE and PU-PTTPE, using ultrasonication-assisted method have been prepared. A comparative study of PU-PF, PU-PFTPE and PU-PTTPE for detection explosives had been performed, and significant fluorescence quenching was observed with the introduction of PA solutions. The as-prepared PU-PF, PU-PFTPE and PU-PTTPE sensors exhibited a superior sensitivity for PA solutions with different concentrations. Remarkably, PU-PF gave a quenching efficiency of 96.2%, higher than 93.5% for PU-PFTPE and 86.7% for PU-PTTPE at a PA concentration of 180 µg·mL-1 in methanol, which was attributed to the effective energy transfer from the fluorophore (PF) to the nitro explosive (PA). This suggested that some ACQ polymers, applied to detect explosives, could afford better performances than AIE or AEE polymers through modification of structures and selection of adequate carriers. At the same time, these chemical sensors can be recycled many times.
Sigma-Aldrich
2-(Tributylstannyl)thiophene, 97%
Sigma-Aldrich
9,9-Dioctyl-2,7-dibromofluorene, 96%
Sigma-Aldrich
9,9-Dioctylfluorene-2,7-diboronic acid, 96%
Sigma-Aldrich
2,5-Bis(tributylstannyl)thiophene, 97%