Merck
  • Crosslinked Polymer Ionic Liquid/Ionic Liquid Blends Prepared by Photopolymerization as Solid-State Electrolytes in Supercapacitors.
Crosslinked Polymer Ionic Liquid/Ionic Liquid Blends Prepared by Photopolymerization as Solid-State Electrolytes in Supercapacitors.
Nanomaterials (Basel, Switzerland) (2018-04-13)
Po-Hsin Wang, Tzong-Liu Wang, Wen-Churng Lin, Hung-Yin Lin, Mei-Hwa Lee, Chien-Hsin Yang
ABSTRACT
A photopolymerization method is used to prepare a mixture of polymer ionic liquid (PIL) and ionic liquid (IL). This mixture is used as a solid-state electrolyte in carbon nanoparticle (CNP)-based symmetric supercapacitors. The solid electrolyte is a binary mixture of a PIL and its corresponding IL. The PIL matrix is a cross-linked polyelectrolyte with an imidazole salt cation coupled with two anions of Br- in PIL-M-(Br) and TFSI- in PIL-M-(TFSI), respectively. The corresponding ionic liquids have imidazolium salt cation coupled with two anions of Br- and TFSI-, respectively. This study investigates the electrochemical characteristics of PILs and their corresponding IL mixtures used as a solid electrolyte in supercapacitors. Results show that a specific capacitance, maximum power density and energy density of 87 and 58 F·g-¹, 40 and 48 kW·kg-¹, and 107 and 59.9 Wh·kg-¹ were achieved in supercapacitors based on (PIL-M-(Br)) and (PIL-M-(TFSI)) solid electrolytes, respectively.
Sigma-Aldrich
Natriuretic Peptide, C-Type, ≥95% (HPLC)
Sigma-Aldrich
1-Vinylimidazole, ≥99%
Sigma-Aldrich
2-Methyl-4′-(methylthio)-2-morpholinopropiophenone, 98%
Sigma-Aldrich
Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide, 97%
Sigma-Aldrich
1,4-Dibromobutane, 99%