
Zinc sulfate

Synonym(s):
Zinc sulfate monohydrate
Linear Formula:
ZnSO4 · H2O
CAS Number:
Molecular Weight:
179.47
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing