
Ursolic acid

Synonym(s):
3β-Hydroxy-12-ursen-28-ic acid, Ursolic Acid
Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing