
Tryptic Soy Agar - Dehydrated Culture Media

Synonym(s):
CASO Agar, Soybean Casein digest Agar, TSA, Tryptone Soya Agar
Products
Product Category
Brand
Color
pH Value
Physical Form
Product Line
Agency
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Package Size
Sterility
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing