Skip to Content
Merck
Tris(triphenylphosphine)rhodium(I) carbonyl hydride 97%

Tris(triphenylphosphine)rhodium(I) carbonyl hydride

Tris(triphenylphosphine)rhodium(I) carbonyl hydride 97%
Synonym(s):
Carbonyltris(triphenylphosphine)rhodium(I) hydride, Hydridocarbonyl-tris(triphenylphosphine)-rhodium(I), Rhodium(I) tris(triphenylphosphine) carbonyl hydride
Linear Formula:
[(C6H5)3P]3Rh(CO)H
CAS Number:
Molecular Weight:
918.78
EC Number:

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing