
Tris(diethylamido)(tert-butylimido)tantalum(V)

Synonym(s):
Tris(diethylamino)(tert-butylimino)tantalum(V)
Linear Formula:
(CH3)3CNTa(N(C2H5)2)3
CAS Number:
Molecular Weight:
468.46
Products
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2