
Tetra-n-butylammonium bromide

Synonym(s):
Tetra-n-butylammonium bromide
Linear Formula:
(CH3CH2CH2CH2)4NBr
CAS Number:
Molecular Weight:
322.37
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing