![Telmisartan for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/385/341/43f3e851-36ba-48b4-9f4d-6b6809ed9877/640/43f3e851-36ba-48b4-9f4d-6b6809ed9877.png)
Telmisartan for peak identification
![Telmisartan for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/385/341/43f3e851-36ba-48b4-9f4d-6b6809ed9877/640/43f3e851-36ba-48b4-9f4d-6b6809ed9877.png)
Synonym(s):
Telmisartan, 4′[(1,4′-Dimethyl-2′-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl][1,1′-biphenyl]-2-carboxylic acid, BIBR 277
Empirical Formula (Hill Notation):
C33H30N4O2
CAS Number:
Molecular Weight:
514.62
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing