
Selenocystamine dihydrochloride

Empirical Formula (Hill Notation):
C4H12N2Se2 · 2HCl
CAS Number:
Molecular Weight:
318.99
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing