Skip to Content
Merck
sec-Butylamine 99%

sec-Butylamine

sec-Butylamine 99%
Synonym(s):
(±)-sec-Butylamine, (±)-2-Aminobutane
Linear Formula:
CH3CH2CH(NH2)CH3
CAS Number:
Molecular Weight:
73.14
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing