
Residual Solvent Class 2 - Methylene chloride

Synonym(s):
Dichloromethane solution, Methylene chloride solution
Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing