
(R)-(−)-1,2-Propanediol

Synonym(s):
(R)-(−)-Propylene glycerol, (R)-(−)-Propylene glycol
Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing