Praziquantel
Synonym(s):
2-(Cyclohexylcarbonyl)-1,2,3,6,7-11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one, Praziquantel
Empirical Formula (Hill Notation):
C19H24N2O2
CAS Number:
Molecular Weight:
312.41
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing