
O-tert-Butylhydroxylamine hydrochloride

Synonym(s):
2-Aminooxy-2-methylpropane hydrochloride, O-(1,1-Dimethylethyl)hydroxylamine hydrochloride, tert-Butoxyamine hydrochloride
Linear Formula:
(CH3)3CONH2 · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing