Skip to Content
Merck
Octanoic-d15 acid ≥98 atom % D, ≥99% (CP)

Octanoic-d15 acid

Octanoic-d15 acid ≥98 atom % D, ≥99% (CP)
Synonym(s):
Caprylic-d15 acid
Linear Formula:
CD3(CD2)6CO2H
CAS Number:
Molecular Weight:
159.30

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing