
Nodularin

Synonym(s):
1,4,8,11,15-Pentaazacyclononadecane, cyclic peptide
Empirical Formula (Hill Notation):
C41H60N8O10
CAS Number:
Molecular Weight:
824.96
Products
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing