
n-Octylglucoside

Synonym(s):
Octyl β-D-glucopyranoside, n-Octyl glucoside, OGP
Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
Products
Product Category
Formula Weight
Manufacturer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing