
Nicotine Related Compound F

Synonym(s):
(±)-Nornicotine, 2-(3-Pyridyl)pyrrolidine, 3-(2-Pyrrolidinyl)pyridine
Empirical Formula (Hill Notation):
C9H12N2
CAS Number:
Molecular Weight:
148.20
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2