
Nicotine ditartrate dihydrate

Synonym(s):
(−)-Nicotine hydrogen tartrate salt dihydrate
Empirical Formula (Hill Notation):
C10H14N2 · 2C4H6O6 · 2H2O
CAS Number:
Molecular Weight:
498.44
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing