Neutral Red Solution, Buffered
Synonym(s):
Neutral Red solution, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride, Toluylene red
Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing