
Naptalam

Synonym(s):
N-(1-Naphthyl)phthalamidic acid
Empirical Formula (Hill Notation):
C18H13NO3
CAS Number:
Molecular Weight:
291.30
Products
Product Category
Formula Weight
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing