
Monoclonal Anti-POU3F2 antibody produced in mouse

Synonym(s):
BRN2, OCT7, POUF3, Anti-39728, Anti-BRN2, Anti-OTF7, Anti-POU domain, class 3, transcription factor 2, Anti-POUF3
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing