
Methyltrioxorhenium(VII)

Synonym(s):
MTO, Methylrhenium(VII) trioxide
Linear Formula:
CH3ReO3
CAS Number:
Molecular Weight:
249.24
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing