Skip to Content
Merck
Methylbenzethonium chloride

Methylbenzethonium chloride

Methylbenzethonium chloride
Synonym(s):
N,N-Dimethyl-N-(2-[2-(methyl-4-[1,1,3,3-tetramethylbutyl]phenoxy)ethoxy]ethyl)benzylammonium chloride
Linear Formula:
C28H44NO2Cl
CAS Number:
Molecular Weight:
462.11
EC Number:

Products

Product Category
Formula Weight
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing