![Methoxyfenozide PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/290/918/80c1aa52-a885-452c-8aae-16999d82577f/640/80c1aa52-a885-452c-8aae-16999d82577f.png)
Methoxyfenozide
![Methoxyfenozide PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/290/918/80c1aa52-a885-452c-8aae-16999d82577f/640/80c1aa52-a885-452c-8aae-16999d82577f.png)
Synonym(s):
3-Methoxy-2-methylbenzoic acid 2-(3,5-dimethylbenzoyl)-2-tert-butylhydrazide
Empirical Formula (Hill Notation):
C22H28N2O3
CAS Number:
Molecular Weight:
368.47
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2