
Methabenzthiazuron

Synonym(s):
1-(Benzothiazol-2-yl)-1,3-dimethylurea
Empirical Formula (Hill Notation):
C10H11N3OS
CAS Number:
Molecular Weight:
221.28
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing