
Melitracen hydrochloride

Synonym(s):
3-[10,10-Dimethyl-9(10H)-anthrylidene]-N,N-dimethylpropylamine hydrochloride, Melitracen hydrochloride
Empirical Formula (Hill Notation):
C21H25N · HCl
CAS Number:
Molecular Weight:
327.89
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing