
Malvidin chloride

Synonym(s):
3,4′,5,7-Tetrahydroxy-3′,5′-dimethoxyflavylium chloride, Malvidin, Oenidin chloride, Primulidin chloride, Syringidin chloride
Empirical Formula (Hill Notation):
C17H15ClO7
CAS Number:
Molecular Weight:
366.75
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing