
L-Proline-1-13C

Empirical Formula (Hill Notation):
13CC4H9O2N
CAS Number:
Molecular Weight:
116.12
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing