![L-PHENYLALANINE AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/286/408/35fd0c7a-6130-4d0d-a4ae-8bbf57bedf75/640/35fd0c7a-6130-4d0d-a4ae-8bbf57bedf75.png)
L-Phenylalanine
![L-PHENYLALANINE AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/286/408/35fd0c7a-6130-4d0d-a4ae-8bbf57bedf75/640/35fd0c7a-6130-4d0d-a4ae-8bbf57bedf75.png)
Synonym(s):
(S)-2-Amino-3-phenylpropionic acid, L-Phenylalanine
Linear Formula:
C9H11NO2
CAS Number:
Molecular Weight:
165.19
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Organoleptic Class
Amino Acid
API Family
Reference Material Type
Special Grade
Technique
Showing 1-12 of 12
Compare
Product Number
Description
Pricing