
Lomustine Related Compound C

Synonym(s):
N,N′-Dicyclohexylurea, 1,3-Dicyclohexylurea
Linear Formula:
C6H11NHCONHC6H11
CAS Number:
Molecular Weight:
224.34
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2