
L-Alanine 4-methoxy-β-naphthylamide hydrochloride

Empirical Formula (Hill Notation):
C14H16N2O2 · HCl
CAS Number:
Molecular Weight:
280.75
Products
Product Category
Brand
Formula Weight
Physical Form
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing