
Guggulsterone Z

Synonym(s):
(Z)-Guggulsterone, (17Z)-Pregna-4,17(20)-diene-3,16-dione, 4,17(20)-trans-Pregnadiene-3,16-dione
Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing