Guaifenesin Impurity C
Synonym(s):
1,1′-Oxybis[3-(2-methoxyphenoxy)-2-propanol]
Empirical Formula (Hill Notation):
C20H26O7
Molecular Weight:
378.42
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing