
Gly-His hydrochloride hydrate

Synonym(s):
Glycyl-L-histidine hydrochloride hydrate
Empirical Formula (Hill Notation):
C8H12N4O3 · HCl · xH2O
CAS Number:
Molecular Weight:
248.67 (anhydrous basis)
Products
Brand
Formula Weight
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing