Skip to Content
Merck
Fluxapyroxad PESTANAL®, analytical standard

Fluxapyroxad

Fluxapyroxad PESTANAL®, analytical standard
Synonym(s):
3-(Difluoromethyl)-1-methyl-N-(3′,4′,5′-trifluorobiphenyl-2-yl)pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C18H12F5N3O
CAS Number:
Molecular Weight:
381.30

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique