Skip to Content
Merck
Flucythrinate PESTANAL®, analytical standard, mixture of stereo isomers

Flucythrinate

Flucythrinate PESTANAL®, analytical standard, mixture of stereo isomers
Synonym(s):
α-Cyano-3-phenoxybenzyl 4-difluoromethoxy-α-isopropylphenylacetate
Empirical Formula (Hill Notation):
C26H23F2NO4
CAS Number:
Molecular Weight:
451.46
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique