![Fluconazole British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/674/544/1f135499-8046-47a7-8fe1-a3f6193e1a74/640/1f135499-8046-47a7-8fe1-a3f6193e1a74.png)
Fluconazole
![Fluconazole British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/674/544/1f135499-8046-47a7-8fe1-a3f6193e1a74/640/1f135499-8046-47a7-8fe1-a3f6193e1a74.png)
Synonym(s):
2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol, Fluconazole
Empirical Formula (Hill Notation):
C13H12F2N6O
CAS Number:
Molecular Weight:
306.27
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Activity Spectrum
Agency
Format
Research Area
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing