
Egeline

Synonym(s):
Aegeline, Egeline
Empirical Formula (Hill Notation):
C18H19NO3
CAS Number:
Molecular Weight:
297.35
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing