Skip to Content
BD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(±)-Ketamine hydrochloride
Synonym(s):
(±)-2-(2-Chlorophenyl)-2-(methylamino)cyclohexanone hydrochloride
Empirical Formula (Hill Notation):
C
13
H
16
ClNO · HCl
CAS Number:
1867-66-9
Molecular Weight:
274.19
EC Number:
217-484-6
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Gene Alias
Physical Form
Research Area
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
K2753
(±)-Ketamine hydrochloride,
≥99% (HPLC), solid, NMDA glutamate receptor antagonist
Expand
View Pricing