
Dioleoylglycerol

Synonym(s):
Diolein, Glycerol-1,2- and -1,3-dioleate
Empirical Formula (Hill Notation):
C39H72O5
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Showing 1-1 of 1
Compare
Product Number
Description
Pricing