
Diatrizoic acid

Synonym(s):
Diatrizoic acid dihydrate, 3,5-Bis(acetylamino)-2,4,6-triiodobenzoic acid dihydrate, Amidotrizoic acid dihydrate
Empirical Formula (Hill Notation):
C11H9I3N2O4 · 2H2O
CAS Number:
Molecular Weight:
649.94
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing