![Dexamethasone for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/348/568/69f70b5c-e320-4119-9e89-5447c8e994fb/640/69f70b5c-e320-4119-9e89-5447c8e994fb.png)
Dexamethasone for peak identification
![Dexamethasone for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/348/568/69f70b5c-e320-4119-9e89-5447c8e994fb/640/69f70b5c-e320-4119-9e89-5447c8e994fb.png)
Synonym(s):
Dexamethasone, (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F
Empirical Formula (Hill Notation):
C22H29FO5
CAS Number:
Molecular Weight:
392.46
Products
Biological Source
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing