Skip to Content
Merck
(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate 98%

(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate

(+)-Dimethyl 2,3-O-isopropylidene-D-tartrate 98%
Synonym(s):
(4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, Dimethyl (4S,5S)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate
Empirical Formula (Hill Notation):
C9H14O6
CAS Number:
Molecular Weight:
218.20

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing