
D-(+)-Glucose solution

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Product Line
Analyte Suitability
Quality Segment
Application
Optical Isomer
Sterilization Method
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing