
Cocaine hydrochloride

Synonym(s):
Ecgonine methyl ester benzoate hydrochloride
Empirical Formula (Hill Notation):
C17H21NO4 · HCl
CAS Number:
Molecular Weight:
339.81
Products
Product Category
Brand
Formula Weight
Physical Form
Agency
Research Area
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing