
Chondroitinase AC from Flavobacterium heparinum

Synonym(s):
Chondroitin AC lyase
CAS Number:
Products
Product Category
Brand
Physical Form
Purity
Recombinant Host
Chemical Composition
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing