
Chlorotriisopropoxytitanium(IV) solution

Linear Formula:
[(CH3)2CHO]3TiCl
CAS Number:
Molecular Weight:
260.58
Products
Formula Weight
Physical Form
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing