
Calcein-AM

Synonym(s):
Calcein O,O′-diacetate tetrakis(acetoxymethyl) ester, Calcein-AM
Empirical Formula (Hill Notation):
C46H46N2O23
CAS Number:
Molecular Weight:
994.86
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-3 of 3
Compare
Product Number
Description
Pricing